SY3H3D680E9E Synthonix Corporation

2-(6-Methoxy-1H-indol-3-yl)acetic acid

  • Cas Number : 103986-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54436.html

Safety Information

MSDS https://synthonix.com/54436.html
Availability details for
Quantity
  • Availability
  • Add to Cart