SY3H3D680EA0 Synthonix Corporation

Methyl 2-(cyanomethyl)-3-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 145498-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54437.html

Safety Information

MSDS https://synthonix.com/54437.html
Availability details for
Quantity
  • Availability
  • Add to Cart