SY3H3D680EA2 Synthonix Corporation

Methyl 5-methoxy-1H-indole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 172595-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/54438.html

Safety Information

MSDS https://synthonix.com/54438.html
Availability details for
Quantity
  • Availability
  • Add to Cart