SY3H3D680EB4 Synthonix Corporation

Methyl 1-(4-iodophenyl)cyclopropanecarboxylate

  • MDL number No MDL found
  • Cas Number : 630384-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54456.html

Safety Information

MSDS https://synthonix.com/54456.html
Availability details for
Quantity
  • Availability
  • Add to Cart