SY3H3D680EB8 Synthonix Corporation

(3-Methoxynaphthalen-1-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 219834-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/54458.html

Safety Information

MSDS https://synthonix.com/54458.html
Availability details for
Quantity
  • Availability
  • Add to Cart