SY3H3D680EBE Synthonix Corporation

Methyl 6-fluorochroman-2-carboxylate

  • Cas Number : 874649-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54465.html

Safety Information

MSDS https://synthonix.com/54465.html
Availability details for
Quantity
  • Availability
  • Add to Cart