SY3H3D680EC1 Synthonix Corporation

(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol

  • Cas Number : 914349-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54486.html

Safety Information

MSDS https://synthonix.com/54486.html
Availability details for
Quantity
  • Availability
  • Add to Cart