SY3H3D680EC4 Synthonix Corporation

4-Morpholin-4-yl-2-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 404010-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54503.html

Safety Information

MSDS https://synthonix.com/54503.html
Availability details for
Quantity
  • Availability
  • Add to Cart