SY3H3D680EC5 Synthonix Corporation

Methyl 4-((2-methoxy-2-oxoethyl)amino)-3-nitrobenzoate

  • Cas Number : 698985-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54504.html

Safety Information

MSDS https://synthonix.com/54504.html
Availability details for
Quantity
  • Availability
  • Add to Cart