SY3H3D680EC7 Synthonix Corporation

Methyl 4-acetamido-5-iodo-2-methoxybenzoate

  • MDL number MFCD13192093
  • Cas Number : 201214-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54532.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54532.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart