SY3H3D680ED3 Synthonix Corporation

5-Methoxy-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 116854-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54581.html

Safety Information

MSDS https://synthonix.com/54581.html
Availability details for
Quantity
  • Availability
  • Add to Cart