SY3H3D680ED4 Synthonix Corporation

5-Methoxychroman-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 112904-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54590.html

Safety Information

MSDS https://synthonix.com/54590.html
Availability details for
Quantity
  • Availability
  • Add to Cart