SY3H3D680ED5 Synthonix Corporation

6-Methoxychroman-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 99199-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54591.html

Safety Information

MSDS https://synthonix.com/54591.html
Availability details for
Quantity
  • Availability
  • Add to Cart