SY3H3D680F20 Synthonix Corporation

Methyl 3-(3-Methoxyphenyl)-3-oxopropionate

  • MDL number No MDL found
  • Cas Number : 779-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/54593.html

Safety Information

MSDS https://synthonix.com/54593.html
Availability details for
Quantity
  • Availability
  • Add to Cart