SY3H3D680F22 Synthonix Corporation

4-((2-Methyl-1H-imidazol-1-yl)methyl)aniline

  • Cas Number : 772311-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54619.html

Safety Information

MSDS https://synthonix.com/54619.html
Availability details for
Quantity
  • Availability
  • Add to Cart