SY3H3D680F24 Synthonix Corporation

2-Morpholinobenzo[d]thiazol-5-amine

  • Cas Number : 1781932-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54623.html

Safety Information

MSDS https://synthonix.com/54623.html
Availability details for
Quantity
  • Availability
  • Add to Cart