SY3H3D680F27 Synthonix Corporation

6-Methoxy-1-methyl-3,4-dihydroquinolin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 187679-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54646.html

Safety Information

MSDS https://synthonix.com/54646.html
Availability details for
Quantity
  • Availability
  • Add to Cart