SY3H3D680F2C Synthonix Corporation

Methyl 5-cyclopropyl-2-methoxynicotinate

  • MDL number No MDL found
  • Cas Number : 888499-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/54654.html

Safety Information

MSDS https://synthonix.com/54654.html
Availability details for
Quantity
  • Availability
  • Add to Cart