SY3H3D680F30 Synthonix Corporation

Methyl 2-(4-bromophenyl)-2,2-dimethylacetate

  • MDL number No MDL found
  • Cas Number : 154825-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54675.html

Safety Information

MSDS https://synthonix.com/54675.html
Availability details for
Quantity
  • Availability
  • Add to Cart