SY3H3D680F34 Synthonix Corporation

4-Methyl-3-phenylpiperazin-2-one

  • MDL number No MDL found
  • Cas Number : 5368-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54698.html

Safety Information

MSDS https://synthonix.com/54698.html
Availability details for
Quantity
  • Availability
  • Add to Cart