SY3H3D680F36 Synthonix Corporation

Methyl 5-((tert-butoxycarbonyl)amino)-4-nitrothiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1351238-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54719.html

Safety Information

MSDS https://synthonix.com/54719.html
Availability details for
Quantity
  • Availability
  • Add to Cart