SY3H3D680F39 Synthonix Corporation

(S)-6-(3-Methylpiperazin-1-yl)nicotinonitrile

  • MDL number No MDL found
  • Cas Number : 1057682-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54723.html

Safety Information

MSDS https://synthonix.com/54723.html
Availability details for
Quantity
  • Availability
  • Add to Cart