SY3H3D680F3B Synthonix Corporation

3-Methyl-3-phenylbutanal

  • Cas Number : 6325-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54755.html

Safety Information

MSDS https://synthonix.com/54755.html
Availability details for
Quantity
  • Availability
  • Add to Cart