SY3H3D680F40 Synthonix Corporation

Methyl 2-methyl-2-phenylpropanoate

  • MDL number No MDL found
  • Cas Number : 57625-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/54778.html

Safety Information

MSDS https://synthonix.com/54778.html
Availability details for
Quantity
  • Availability
  • Add to Cart