SY3H3D680F4B Synthonix Corporation

(R)-5-Methyl-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number No MDL found
  • Cas Number : 1055321-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54825.html

Safety Information

MSDS https://synthonix.com/54825.html
Availability details for
Quantity
  • Availability
  • Add to Cart