SY3H3D680F4E Synthonix Corporation

3-Morpholinobenzyl Alcohol

  • MDL number No MDL found
  • Cas Number : 145127-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54862.html

Safety Information

MSDS https://synthonix.com/54862.html
Availability details for
Quantity
  • Availability
  • Add to Cart