SY3H3D680F52 Synthonix Corporation

Methyl 1,4,5,6,7,8-hexahydrocyclohepta[b]pyrrole-2-carboxylate

  • Cas Number : 100445-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54876.html

Safety Information

MSDS https://synthonix.com/54876.html
Availability details for
Quantity
  • Availability
  • Add to Cart