SY3H3D680F56 Synthonix Corporation

2-(2-(Methylthio)phenyl)pyrrolidine

  • Cas Number : 383127-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54904.html

Safety Information

MSDS https://synthonix.com/54904.html
Availability details for
Quantity
  • Availability
  • Add to Cart