SY3H3D680F5A Synthonix Corporation

1-(4-(Methylsulfonyl)phenyl)piperazine

  • MDL number No MDL found
  • Cas Number : 187669-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54927.html

Safety Information

MSDS https://synthonix.com/54927.html
Availability details for
Quantity
  • Availability
  • Add to Cart