SY3H3D680F5C Synthonix Corporation

(2-(4-(Methoxycarbonyl)piperidin-1-yl)pyrimidin-5-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1536398-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54937.html

Safety Information

MSDS https://synthonix.com/54937.html
Availability details for
Quantity
  • Availability
  • Add to Cart