SY3H3D680F60 Synthonix Corporation

(3-Methyl-4-morpholinophenyl)boronic acid

  • Cas Number : 1426245-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54944.html

Safety Information

MSDS https://synthonix.com/54944.html
Availability details for
Quantity
  • Availability
  • Add to Cart