SY3H3D680F61 Synthonix Corporation

2-Mesitylethanol

  • MDL number MFCD00020609
  • Cas Number : 6950-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54948.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54948.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart