SY3H3D680F64 Synthonix Corporation

1-(4-Methoxyphenyl)-2-methylpropan-1-ol

  • MDL number MFCD11934907
  • Cas Number : 18228-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54959.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54959.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart