SY3H3D680F65 Synthonix Corporation

3-((4-Methoxybenzyl)oxy)propan-1-ol

  • MDL number No MDL found
  • Cas Number : 135362-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54961.html

Safety Information

MSDS https://synthonix.com/54961.html
Availability details for
Quantity
  • Availability
  • Add to Cart