SY3H3D680F6B Synthonix Corporation

2-(Methyl(2-phenoxyethyl)amino)ethanol

  • Cas Number : 113824-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54994.html

Safety Information

MSDS https://synthonix.com/54994.html
Availability details for
Quantity
  • Availability
  • Add to Cart