SY3H3D680F6F Synthonix Corporation

2-Methyladamantan-2-ol

  • MDL number MFCD00074811
  • Cas Number : 702-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55019.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55019.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart