SY3H3D680F74 Synthonix Corporation

malonic acid dibutyl ester

  • Cas Number : 1190-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55093.html

Safety Information

MSDS https://synthonix.com/55093.html
Availability details for
Quantity
  • Availability
  • Add to Cart