SY3H3D680F76 Synthonix Corporation

(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine

  • MDL number No MDL found
  • Cas Number : 155726-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55122.html

Safety Information

MSDS https://synthonix.com/55122.html
Availability details for
Quantity
  • Availability
  • Add to Cart