SY3H3D680F7A Synthonix Corporation

Methyl 6-cyano-1H-indole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 104291-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55239.html

Safety Information

MSDS https://synthonix.com/55239.html
Availability details for
Quantity
  • Availability
  • Add to Cart