SY3H3D680F84 Synthonix Corporation

Methyl 8-bromoquinoline-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 253787-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55271.html

Safety Information

MSDS https://synthonix.com/55271.html
Availability details for
Quantity
  • Availability
  • Add to Cart