SY3H3D680F87 Synthonix Corporation

Methyl 4-chloroquinoline-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 178984-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55276.html

Safety Information

MSDS https://synthonix.com/55276.html
Availability details for
Quantity
  • Availability
  • Add to Cart