SY3H3D680F8A Synthonix Corporation

Methyl 5-(3-bromophenyl)isoxazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 745078-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55283.html

Safety Information

MSDS https://synthonix.com/55283.html
Availability details for
Quantity
  • Availability
  • Add to Cart