SY3H3D680F90 Synthonix Corporation

5-(2-Methyl-1H-imidazol-1-yl)-2-nitrobenzoic acid

  • Cas Number : 954265-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55335.html

Safety Information

MSDS https://synthonix.com/55335.html
Availability details for
Quantity
  • Availability
  • Add to Cart