SY3H3D680F91 Synthonix Corporation

Methyl 3-(2-nitrophenyl)-1H-pyrazole-5-carboxylate

  • Cas Number : 57446-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55336.html

Safety Information

MSDS https://synthonix.com/55336.html
Availability details for
Quantity
  • Availability
  • Add to Cart