SY3H3D680F99 Synthonix Corporation

8-Methoxyquinoline-4-carbaldehyde

  • Cas Number : 103854-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55363.html

Safety Information

MSDS https://synthonix.com/55363.html
Availability details for
Quantity
  • Availability
  • Add to Cart