SY3H3D680F9C Synthonix Corporation

6-Methylquinoline-8-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 55706-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55366.html

Safety Information

MSDS https://synthonix.com/55366.html
Availability details for
Quantity
  • Availability
  • Add to Cart