SY3H3D680F9E Synthonix Corporation

1-Methoxy-4-isoquinolinecarboxaldehyde

  • MDL number No MDL found
  • Cas Number : 1005772-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55368.html

Safety Information

MSDS https://synthonix.com/55368.html
Availability details for
Quantity
  • Availability
  • Add to Cart