SY3H3D680F9F Synthonix Corporation

2-(4-Methoxyphenyl)thiazole-5-carbaldehyde

  • Cas Number : 914348-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55376.html

Safety Information

MSDS https://synthonix.com/55376.html
Availability details for
Quantity
  • Availability
  • Add to Cart