SY3H3D680FA4 Synthonix Corporation

Methyl 2-formyl-1H-indole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 880360-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55388.html

Safety Information

MSDS https://synthonix.com/55388.html
Availability details for
Quantity
  • Availability
  • Add to Cart