SY3H3D680FA6 Synthonix Corporation

5-Methyl-2-phenyloxazole-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 18735-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/55391.html

Safety Information

MSDS https://synthonix.com/55391.html
Availability details for
Quantity
  • Availability
  • Add to Cart